Assay Design
Gene Transcripts
- Gene Symbol
- BGAL11
- Entrez Gene ID
-
829653
- Gene Name
- beta-galactosidase 11
- Gene Aliases
-
AT4G35010, BETA-GALACTOSIDASE, M4E13.70, M4E13_70, beta-galactosidase 11
- Chromosome Location
-
Chr.4: 16667837 - 16672188 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_119667.3 |
NP_567973.1 |
9 - 10 |
1335 |
70 |
GenBank mRNA |
AJ270307.1 |
- |
9 - 10 |
1224 |
70 |
View Full Product Details