Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001335615.1 | NP_001325303.1 | 8 - 9 | 945 | 88 |
NM_179653.4 | NP_849984.1 | 8 - 9 | 1066 | 88 | |
GenBank mRNA | AF324344.1 | - | 8 - 9 | 888 | 88 |
AY063893.1 | - | 8 - 9 | 918 | 88 | |
AY089963.1 | - | 8 - 9 | 774 | 88 | |
AY113993.1 | - | 8 - 9 | 774 | 88 | |
GU198365.1 | - | 8 - 9 | 852 | 88 |