Assay Design
Gene Transcripts
- Gene Symbol
- ASHR2
- Entrez Gene ID
-
816483
- Gene Name
- ASH1-related protein 2
- Gene Aliases
-
ASH1-related 2, ASH1-related protein 2, AT2G19640, F3P11.24, F3P11_24, SDG39, SET DOMAIN PROTEIN 39
- Chromosome Location
-
Chr.2: 8491192 - 8492920 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_179660.2 |
NP_849991.1 |
1 - 1 |
1252 |
85 |
GenBank mRNA |
BT004010.1 |
- |
- |
1816 |
85 |
View Full Product Details