Assay Design
Gene Transcripts
- Gene Symbol
- BGLU14
- Entrez Gene ID
-
817104
- Gene Name
- beta glucosidase 14
- Gene Aliases
-
AT2G25630, F3N11.8, F3N11_8, beta glucosidase 14
- Chromosome Location
-
Chr.2: 10908360 - 10909880 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_179734.1 |
NP_850065.1 |
1 - 2 |
1123 |
152 |
View Full Product Details