Assay Design



Gene Transcripts

Gene Symbol
YAB5
Entrez Gene ID
817199
Gene Name
plant-specific transcription factor YABBY family protein
Gene Aliases
AT2G26580, T9J22.25, T9J22_25, YABBY5
Chromosome Location
Chr.2: 11303455 - 11307010 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_179749.3 NP_850080.1 6 - 7 776 88
NM_179750.3 NP_850081.1 7 - 8 619 88
GenBank mRNA AK119091.1 - 6 - 7 545 88

View Full Product Details