Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001036351.2 | NP_001031428.1 | 3 - 4 | 2121 | 110 |
NM_128269.4 | NP_565636.1 | 2 - 3 | 2418 | 110 | |
NM_179767.2 | NP_850098.1 | 1 - 2 | 2565 | 110 | |
GenBank mRNA | AK317350.1 | - | 3 - 4 | 2064 | 110 |
AY057491.1 | - | 2 - 3 | 2356 | 110 | |
BT002690.1 | - | 1 - 2 | 2153 | 110 |