Assay Design



Gene Transcripts

Gene Symbol
PIFI
Entrez Gene ID
820827
Gene Name
post-illumination chlorophyll fluorescence increase
Gene Aliases
AT3G15840, post-illumination chlorophyll fluorescence increase
Chromosome Location
Chr.3: 5356587 - 5358596 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035629.1 NP_001030706.1 2 - 3 209 74
NM_001035630.1 NP_001030707.1 2 - 3 267 74
NM_001202975.1 NP_001189904.1 2 - 3 377 74
NM_112454.3 NP_566528.1 2 - 3 364 74
GenBank mRNA AY035025.1 - 2 - 3 223 74
AY133879.1 - 2 - 3 186 74

View Full Product Details