Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001035629.1 | NP_001030706.1 | 5 - 6 | 477 | 89 |
NM_001035630.1 | NP_001030707.1 | 5 - 6 | 544 | 89 | |
NM_001125170.1 | NP_001118642.1 | 4 - 5 | 565 | 89 | |
NM_001202975.1 | NP_001189904.1 | 5 - 6 | 654 | 89 | |
NM_112454.3 | NP_566528.1 | 5 - 6 | 641 | 89 | |
GenBank mRNA | AY035025.1 | - | 5 - 6 | 500 | 89 |
AY133879.1 | - | 5 - 6 | 463 | 89 |