Assay Design



Gene Transcripts

Gene Symbol
NAI2
Entrez Gene ID
820839
Gene Name
DNA topoisomerase-like protein
Gene Aliases
AT3G15950
Chromosome Location
Chr.3: 5397541 - 5402653 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035631.2 NP_001030708.1 15 - 16 985 105
NM_001338192.1 NP_001326807.1 9 - 10 637 105
NM_112465.4 NP_188216.2 17 - 18 1099 105
GenBank mRNA AY120750.1 - 17 - 18 1098 105
BT001207.1 - 17 - 18 1057 105

View Full Product Details