Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001035631.2 | NP_001030708.1 | 1 - 2 | 111 | 129 |
NM_001338191.1 | NP_001326809.1 | 1 - 2 | 111 | 129 | |
NM_001338192.1 | NP_001326807.1 | 1 - 2 | 111 | 129 | |
NM_001338193.1 | NP_001326808.1 | 1 - 2 | 112 | 129 | |
NM_112465.4 | NP_188216.2 | 1 - 2 | 111 | 129 | |
GenBank mRNA | AY120750.1 | - | 1 - 2 | 110 | 129 |
BT001207.1 | - | 1 - 2 | 69 | 129 |