Assay Design



Gene Transcripts

Gene Symbol
NAI2
Entrez Gene ID
820839
Gene Name
DNA topoisomerase-like protein
Gene Aliases
AT3G15950
Chromosome Location
Chr.3: 5397541 - 5402653 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035631.2 NP_001030708.1 22 - 23 1342 95
NM_001338191.1 NP_001326809.1 16 - 17 1015 95
NM_001338192.1 NP_001326807.1 16 - 17 994 95
NM_001338193.1 NP_001326808.1 14 - 15 881 95
NM_112465.4 NP_188216.2 24 - 25 1456 95
GenBank mRNA AY120750.1 - 24 - 25 1455 95
BT001207.1 - 24 - 25 1414 95

View Full Product Details