Assay Design
Gene Transcripts
- Gene Symbol
- DEG12
- Entrez Gene ID
-
820903
- Gene Name
- protease Do-like protein
- Gene Aliases
-
AT3G16550, DEGP protease 12, DEGP12, degradation of periplasmic proteins 12
- Chromosome Location
-
Chr.3: 5629920 - 5632441 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_112527.2 |
NP_566552.2 |
6 - 7 |
1250 |
147 |
View Full Product Details