Assay Design



Gene Transcripts

Gene Symbol
TCTP
Entrez Gene ID
820916
Gene Name
translationally controlled tumor protein
Gene Aliases
AT3G16640, translationally controlled tumor protein
Chromosome Location
Chr.3: 5669317 - 5670842 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112537.4 NP_188286.1 3 - 4 507 61
GenBank mRNA AF215897.1 - 3 - 4 444 61
AF361815.1 - 3 - 4 480 61
AY045802.1 - 3 - 4 474 61
AY056065.1 - 3 - 4 394 61
AY072337.1 - 3 - 4 479 61
AY079333.1 - 3 - 4 394 61
AY114601.1 - 3 - 4 394 61

View Full Product Details