Assay Design
Gene Transcripts
- Gene Symbol
- PUM24
- Entrez Gene ID
-
820934
- Gene Name
- pumilio 24
- Gene Aliases
-
APUM24, AT3G16810, pumilio 24
- Chromosome Location
-
Chr.3: 5723222 - 5727664 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_112555.2 |
NP_566559.1 |
11 - 12 |
1243 |
94 |
GenBank mRNA |
AY045638.1 |
- |
11 - 12 |
1200 |
94 |
View Full Product Details