Assay Design
Gene Transcripts
- Gene Symbol
- PMEI2
- Entrez Gene ID
-
820981
- Gene Name
- pectin methylesterase inhibitor 2
- Gene Aliases
-
AT3G17220, ATPMEI2, pectin methylesterase inhibitor 2
- Chromosome Location
-
Chr.3: 5883328 - 5884191 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_112599.2 |
NP_188348.1 |
1 - 1 |
776 |
122 |
- Gene Symbol
- HS1
- Entrez Gene ID
-
820980
- Gene Name
- heat stable protein 1
- Gene Aliases
-
A. THALIANA HEAT STABLE PROTEIN 1, AT3G17210, ATHS1, heat stable protein 1
- Chromosome Location
-
Chr.3: 5881919 - 5883200 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK230465.1 |
- |
2 - 2 |
886 |
122 |
View Full Product Details