Assay Design



Gene Transcripts

Gene Symbol
AGD15
Entrez Gene ID
821033
Gene Name
ARF-GAP domain 15
Gene Aliases
ARF-GAP domain 15, AT3G17660
Chromosome Location
Chr.3: 6037598 - 6039407 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001338298.1 NP_001327025.1 2 - 3 230 101
NM_001338299.1 NP_001327028.1 2 - 3 230 101
NM_001338300.1 NP_001327026.1 2 - 3 230 101
NM_001338301.1 NP_001327027.1 1 - 2 117 101
NM_112647.4 NP_188393.2 2 - 3 230 101
GenBank mRNA AK228639.1 - 2 - 3 230 101

View Full Product Details