Assay Design



Gene Transcripts

Gene Symbol
CNGC19
Entrez Gene ID
821036
Gene Name
cyclic nucleotide gated channel 19
Gene Aliases
AT3G17690, ATCNGC19, CYCLIC NUCLEOTIDE-GATED CHANNEL 19, cyclic nucleotide gated channel 19
Chromosome Location
Chr.3: 6045002 - 6048529 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001338305.1 NP_001325958.1 5 - 6 1692 139
NM_001338306.1 NP_001325957.1 5 - 6 1692 139
NM_112650.4 NP_188396.2 5 - 6 1692 139

View Full Product Details