Assay Design



Gene Transcripts

Gene Symbol
UNG
Entrez Gene ID
821394
Gene Name
uracil dna glycosylase
Gene Aliases
AT3G18630, ATUNG, uracil dna glycosylase
Chromosome Location
Chr.3: 6410958 - 6413196 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112749.5 NP_188493.1 2 - 3 708 105
GenBank mRNA AY084956.1 - 2 - 3 580 105
BT029175.1 - 2 - 3 514 105

View Full Product Details