Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_112912.3 | NP_188656.1 | 5 - 6 | 931 | 82 |
GenBank mRNA | AB105106.1 | - | 5 - 6 | 832 | 82 |
AF521661.1 | - | 5 - 6 | 835 | 82 | |
AY120765.1 | - | 5 - 6 | 858 | 82 | |
BT000949.1 | - | 5 - 6 | 796 | 82 |