Assay Design



Gene Transcripts

Gene Symbol
PUM5
Entrez Gene ID
821569
Gene Name
pumilio 5
Gene Aliases
APUM5, AT3G20250, pumilio 5
Chromosome Location
Chr.3: 7058871 - 7063372 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001338465.1 NP_001319600.1 8 - 9 2595 69
NM_001338466.1 NP_001327305.1 7 - 8 2703 69
NM_001338467.1 NP_001327304.1 7 - 8 2779 69

View Full Product Details