Assay Design



Gene Transcripts

Gene Symbol
UBC17
Entrez Gene ID
829793
Gene Name
ubiquitin-conjugating enzyme 17
Gene Aliases
AP22.89, AP22_89, AT4G36410, ubiquitin-conjugating enzyme 17
Chromosome Location
Chr.4: 17201922 - 17202988 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119804.2 NP_568004.1 2 - 3 181 83
GenBank mRNA AF028340.1 - 2 - 3 152 83
AY088076.1 - 2 - 3 176 83
DQ027031.1 - 2 - 3 134 83

View Full Product Details