Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001036721.2 | NP_001031798.1 | 2 - 3 | 321 | 84 |
NM_119864.5 | NP_195418.1 | 1 - 2 | 229 | 84 | |
GenBank mRNA | AY363869.1 | - | 1 - 2 | 123 | 84 |
BT024826.1 | - | 1 - 2 | 123 | 84 |