Assay Design
Gene Transcripts
- Gene Symbol
- HMA1
- Entrez Gene ID
-
829881
- Gene Name
- heavy metal atpase 1
- Gene Aliases
-
AP22.4, AP22_4, ARABIDOPSIS THALIANA HEAVY METAL ATPASE 1, AT4G37270, ATHMA1, heavy metal atpase 1
- Chromosome Location
-
Chr.4: 17541771 - 17546546 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_119890.7 |
NP_195444.1 |
11 - 12 |
2403 |
72 |
GenBank mRNA |
AJ872069.1 |
- |
11 - 12 |
2235 |
72 |
View Full Product Details