Assay Design
Gene Transcripts
- Gene Symbol
- PSK6
- Entrez Gene ID
-
829928
- Gene Name
- phytosulfokine 6 precursor
- Gene Aliases
-
AT4G37720, ATPSK6, T28I19.1, phytosulfokine 6 precursor
- Chromosome Location
-
Chr.4: 17718953 - 17719321 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_119934.2 |
NP_195486.1 |
1 - 2 |
133 |
63 |
View Full Product Details