Assay Design



Gene Transcripts

Gene Symbol
CSLB03
Entrez Gene ID
817814
Gene Name
cellulose synthase-like B3
Gene Aliases
AT2G32530, ATCSLB03, ATCSLB3, T26B15.9, T26B15_9, cellulose synthase-like B3
Chromosome Location
Chr.2: 13809019 - 13813572 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_179859.4 NP_850190.1 2 - 3 846 80
GenBank mRNA AY090243.1 - 2 - 3 637 80
BT002290.1 - 2 - 3 582 80
KJ138881.1 - 2 - 3 582 80

View Full Product Details