Assay Design



Gene Transcripts

Gene Symbol
HSP60-2
Entrez Gene ID
817883
Gene Name
heat shock protein 60-2
Gene Aliases
AT2G33210, F25I18.5, F25I18_5, heat shock protein 60-2
Chromosome Location
Chr.2: 14074767 - 14078772 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202736.1 NP_001189665.1 11 - 12 1303 97
NM_179872.3 NP_850203.1 11 - 12 1346 97
GenBank mRNA AK175415.1 - 11 - 12 1305 97
AY136360.1 - 11 - 12 1182 97
BT000103.1 - 11 - 12 1142 97

View Full Product Details