Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001202736.1 | NP_001189665.1 | 12 - 13 | 1397 | 91 |
NM_179872.3 | NP_850203.1 | 12 - 13 | 1440 | 91 | |
GenBank mRNA | AK175415.1 | - | 12 - 13 | 1399 | 91 |
AY136360.1 | - | 12 - 13 | 1276 | 91 | |
BT000103.1 | - | 12 - 13 | 1236 | 91 |