Assay Design



Gene Transcripts

Gene Symbol
AT2G34150
Entrez Gene ID
817976
Gene Name
SCAR family protein
Gene Aliases
AT2G34150, ATRANGAP2, ATSCAR1, SCAR1, T14G11.27, T14G11_27, WAVE1, WISKOTT-ALDRICH SYNDROME PROTEIN FAMILY VERPROLIN HOMOLOGOUS PROTEIN 1
Chromosome Location
Chr.2: 14419263 - 14423478 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036397.3 NP_001031474.1 8 - 9 2618 68
NM_001336486.1 NP_001324986.1 5 - 6 2252 68
NM_001336487.1 NP_001324987.1 7 - 8 2295 68
NM_001336488.1 NP_001324988.1 6 - 7 2291 68
NM_179894.2 NP_850225.1 8 - 9 2644 68

View Full Product Details