Assay Design



Gene Transcripts

Gene Symbol
SNZ
Entrez Gene ID
818510
Gene Name
Integrase-type DNA-binding superfamily protein
Gene Aliases
AT2G39250, SCHNARCHZAPFEN, T16B24.11, T16B24_11
Chromosome Location
Chr.2: 16388691 - 16391176 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336762.1 NP_001325386.1 4 - 5 663 104
NM_179982.3 NP_850313.2 4 - 5 766 104
GenBank mRNA AY576276.1 - 4 - 5 521 104

View Full Product Details