Assay Design



Gene Transcripts

Gene Symbol
UGT72B3
Entrez Gene ID
837503
Gene Name
UDP-glucosyl transferase 72B3
Gene Aliases
AT1G01420, F6F3.22, F6F3_22, UDP-glucosyl transferase 72B3
Chromosome Location
Chr.1: 154367 - 156178 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001331274.1 NP_001322549.1 1 - 1 918 84
NM_100024.2 NP_171649.1 1 - 1 751 84
GenBank mRNA BT030469.1 - 1 - 1 751 84
KJ139036.1 - 1 - 1 751 84

View Full Product Details