Assay Design



Gene Transcripts

Gene Symbol
GA2OX6
Entrez Gene ID
839508
Gene Name
gibberellin 2-oxidase 6
Gene Aliases
ARABIDOPSIS THALIANA GIBBERELLIN 2-OXIDASE 4, ARABIDOPSIS THALIANA GIBBERELLIN 2-OXIDASE 6, AT1G02400, ATGA2OX4, ATGA2OX6, Arabidopsis thaliana gibberellin 2-oxidase 4, DOWNSTREAM TARGET OF AGL15 1, DTA1, T6A9.9, T6A9_9, gibberellin 2-oxidase 6
Chromosome Location
Chr.1: 486800 - 489651 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_100121.5 NP_171742.1 2 - 3 908 80
GenBank mRNA AK228830.1 - 2 - 3 807 80
AY859741.1 - 2 - 3 744 80
BT014886.1 - 2 - 3 744 80

View Full Product Details