Assay Design



Gene Transcripts

Gene Symbol
BXL2
Entrez Gene ID
837940
Gene Name
beta-xylosidase 2
Gene Aliases
AT1G02640, ATBXL2, BETA-XYLOSIDASE 2, T14P4.8, T14P4_8, beta-xylosidase 2
Chromosome Location
Chr.1: 564018 - 567771 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_100144.3 NP_563659.1 1 - 2 682 58
GenBank mRNA AF367266.1 - 1 - 2 495 58
AY143952.1 - 1 - 2 407 58

View Full Product Details