Assay Design
Gene Transcripts
- Gene Symbol
- CEL2
- Entrez Gene ID
-
839385
- Gene Name
- cellulase 2
- Gene Aliases
-
4-BETA GLUCANASE, AT1G02800, ATCEL2, ENDO-1, F22D16.21, F22D16_21, cellulase 2
- Chromosome Location
-
Chr.1: 613197 - 616231 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_100159.3 |
NP_171779.1 |
3 - 4 |
785 |
111 |
GenBank mRNA |
AK228979.1 |
- |
3 - 4 |
745 |
111 |
View Full Product Details