Assay Design



Gene Transcripts

Gene Symbol
LPA1
Entrez Gene ID
839350
Gene Name
tetratricopeptide repeat (TPR)-containing protein
Gene Aliases
AT1G02910, F22D16.9, F22D16_9, LOW PSII ACCUMULATION1
Chromosome Location
Chr.1: 655568 - 658216 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001331364.1 NP_001320623.1 3 - 4 572 81
NM_100172.3 NP_171790.1 3 - 4 598 81

View Full Product Details