Assay Design



Gene Transcripts

Gene Symbol
FZL
Entrez Gene ID
839566
Gene Name
FZO-like protein
Gene Aliases
AT1G03160, F10O3.1, F10O3_1, FZO-like
Chromosome Location
Chr.1: 761261 - 766194 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083983.1 NP_001077452.1 5 - 6 1436 56
NM_001331388.1 NP_001322638.1 5 - 6 1457 56
NM_100198.5 NP_171815.3 5 - 6 1457 56
GenBank mRNA AK176297.1 - 5 - 6 1429 56
AK227660.1 - 5 - 6 1429 56
DQ462573.1 - 5 - 6 1436 56

View Full Product Details