Assay Design



Gene Transcripts

Gene Symbol
BSU1
Entrez Gene ID
838804
Gene Name
Serine/threonine protein phosphatase family protein
Gene Aliases
AT1G03445, BRI1 SUPPRESSOR 1, F21B7.7
Chromosome Location
Chr.1: 854410 - 859701 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001331412.1 NP_001321215.1 12 - 13 1279 108
NM_100227.6 NP_171844.6 12 - 13 1279 108
GenBank mRNA AY372269.1 - 12 - 13 1279 108

View Full Product Details