Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001331455.1 | NP_001322970.1 | 2 - 3 | 466 | 105 |
NM_100279.5 | NP_171894.1 | 2 - 3 | 466 | 105 | |
GenBank mRNA | AY044049.1 | - | 2 - 3 | 464 | 105 |
BT029214.1 | - | 2 - 3 | 229 | 105 |