Assay Design



Gene Transcripts

Gene Symbol
PCS2
Entrez Gene ID
839354
Gene Name
phytochelatin synthase 2
Gene Aliases
AT1G03980, ATPCS2, F21M11.9, F21M11_9, phytochelatin synthase 2
Chromosome Location
Chr.1: 1019311 - 1022108 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001331453.1 NP_001318916.1 5 - 6 929 152
NM_001331454.1 NP_001322971.1 6 - 7 1405 152
NM_100279.5 NP_171894.1 7 - 8 1339 152
GenBank mRNA AY044049.1 - 7 - 8 1337 152
BT029214.1 - 7 - 8 1102 152

View Full Product Details