Assay Design



Gene Transcripts

Gene Symbol
IAA10
Entrez Gene ID
839290
Gene Name
indoleacetic acid-induced protein 10
Gene Aliases
AT1G04100, AUXIN-INDUCED PROTEIN 10, F20D22.13, F20D22_13, indoleacetic acid-induced protein 10
Chromosome Location
Chr.1: 1059470 - 1061209 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_100291.3 NP_171906.1 2 - 3 918 125
GenBank mRNA AF332396.1 - 2 - 3 579 125
U18412.1 - 2 - 3 884 125

View Full Product Details