Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001197977.1 | NP_001184906.1 | 11 - 12 | 4879 | 88 |
NM_001331463.1 | NP_001322077.1 | 12 - 13 | 4761 | 88 | |
NM_100293.3 | NP_171908.1 | 11 - 12 | 4927 | 88 | |
GenBank mRNA | AF225908.1 | - | 11 - 12 | 4662 | 88 |
Y11250.1 | - | - | 4642 | 88 |