Assay Design



Gene Transcripts

Gene Symbol
KAB1
Entrez Gene ID
839450
Gene Name
potassium channel beta subunit 1
Gene Aliases
AT1G04690, K+ CHANNEL BETA SUBUNIT PROTEIN, KV-BETA1, POTASSIUM CHANNEL BETA SUBUNIT HOMOLOG, T1G11.6, T1G11_6, potassium channel beta subunit 1
Chromosome Location
Chr.1: 1313443 - 1315823 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_100349.4 NP_171963.1 2 - 3 778 72
GenBank mRNA AF061570.1 - 2 - 3 642 72
AY050821.1 - 2 - 3 612 72
AY091424.1 - 2 - 3 559 72

View Full Product Details