Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001331554.1 | NP_001320882.1 | 1 - 1 | 1737 | 112 |
NM_100435.3 | NP_563742.1 | 1 - 1 | 1449 | 112 | |
GenBank mRNA | AF196777.1 | - | 1 - 1 | 1449 | 112 |
AF367358.1 | - | 1 - 1 | 1449 | 112 |