Assay Design



Gene Transcripts

Gene Symbol
CHX23
Entrez Gene ID
837063
Gene Name
cation/H+ exchanger 23
Gene Aliases
AT1G05580, ATCHX23, F3F20.2, F3F20_2, cation/H+ exchanger 23
Chromosome Location
Chr.1: 1665356 - 1668924 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035898.1 NP_001030975.1 3 - 4 1366 77
NM_100438.5 NP_172049.2 3 - 4 1382 77
GenBank mRNA AY072159.1 - 3 - 4 1360 77
AY096415.1 - 3 - 4 1252 77

View Full Product Details