Assay Design
Gene Transcripts
- Gene Symbol
- MAPKKK13
- Entrez Gene ID
-
837226
- Gene Name
- mitogen-activated protein kinase kinase kinase 13
- Gene Aliases
-
AT1G07150, F10K1.14, F10K1_14, mitogen-activated protein kinase kinase kinase 13
- Chromosome Location
-
Chr.1: 2193941 - 2195996 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_100589.3 |
NP_172195.1 |
1 - 1 |
1665 |
78 |
View Full Product Details