Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_112965.1 | NP_188710.1 | 11 - 12 | 1192 | 82 |
GenBank mRNA | AF129516.1 | - | 11 - 12 | 1192 | 82 |
AK117114.1 | - | 11 - 12 | 1049 | 82 | |
BT005326.1 | - | 11 - 12 | 994 | 82 |