Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_112965.1 | NP_188710.1 | 4 - 5 | 643 | 124 |
GenBank mRNA | AF129516.1 | - | 4 - 5 | 643 | 124 |
AK117114.1 | - | 4 - 5 | 500 | 124 | |
BT005326.1 | - | 4 - 5 | 445 | 124 |