Assay Design



Gene Transcripts

Gene Symbol
PEN2
Entrez Gene ID
819056
Gene Name
Glycosyl hydrolase superfamily protein
Gene Aliases
AT2G44490, BETA GLUCOSIDASE 26, BGLU26, F4I1.30, PENETRATION 2
Chromosome Location
Chr.2: 18364652 - 18367835 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130012.4 NP_181977.1 7 - 8 1061 115
GenBank mRNA AY091016.1 - 7 - 8 930 115
BT000990.1 - 6 - 7 841 115

View Full Product Details