Assay Design



Gene Transcripts

Gene Symbol
PEN2
Entrez Gene ID
819056
Gene Name
Glycosyl hydrolase superfamily protein
Gene Aliases
AT2G44490, BETA GLUCOSIDASE 26, BGLU26, F4I1.30, PENETRATION 2
Chromosome Location
Chr.2: 18364652 - 18367835 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130012.4 NP_181977.1 9 - 10 1415 134
GenBank mRNA AY091016.1 - 9 - 10 1284 134
BT000990.1 - 8 - 9 1195 134

View Full Product Details