Assay Design



Gene Transcripts

Gene Symbol
PVA12
Entrez Gene ID
819122
Gene Name
plant VAP homolog 12
Gene Aliases
AT2G45140, T14P1.5, plant VAP homolog 12
Chromosome Location
Chr.2: 18610806 - 18613302 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130077.3 NP_182039.1 5 - 6 707 80
GenBank mRNA AY088399.1 - 5 - 6 646 80
AY093014.1 - 5 - 6 707 80
BT006283.1 - 5 - 6 484 80

View Full Product Details