Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_130126.4 | NP_566050.1 | 4 - 5 | 412 | 117 |
GenBank mRNA | AF378876.1 | - | 4 - 5 | 367 | 117 |
AK227453.1 | - | 4 - 5 | 363 | 117 | |
AY050474.1 | - | 4 - 5 | 352 | 117 |